4-Cyano-3-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Cyano-3-fluorobenzeneboronic acid

A183122-1G A183122-25G A183122-5G A183122-100G A183122-10G A183122-250MG
103815-610EA 6 USD
103815-610 103846-172 77083-762 77083-764 77083-766 77111-480 77111-482 77111-484
4-Cyano-3-fluorophenylboronic acid 97%
4-Cyano-3-fluorophenylboronic acid

Caution: for Research Use Only

Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
Melting Pt: 362…366 °C
Flash Pt: 168.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03411549
CAS Number: 843663-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR