4-Aminopyrimidine 97%

Supplier: Ambeed
Warning

A145044-100G A145044-10G A145044-250MG A145044-500G A145044-1G A145044-25G A145044-5G
103803-486EA 18 USD
103803-486 103841-568 77109-994 77109-996 77109-998 77110-000 77299-434 77316-838 77316-840
4-Aminopyrimidine 97%
4-Aminopyrimidine

Caution: for Research Use Only

Formula: C₄H₅N₃
MW: 95.1
Boiling Pt: 240.7 °C (760 mm Hg)
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00006112
CAS Number: 591-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR