4-Amino-3-fluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-3-fluorobenzoic Acid
A140966-100G
A140966-10G
A140966-5G
A140966-25G
103802-926EA
8
USD
103802-926
103833-956
77137-306
77137-308
77299-178
77316-052
4-Amino-3-fluorobenzoic acid 98%
4-Amino-3-fluorobenzoic acid
Caution: for Research Use Only
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 320.8 °C (760 mm Hg) Melting Pt: 215…218 °C Flash Pt: 147.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD01660374 CAS Number: 455-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...