4-Amino-α,α,α-trifluoro-o-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-(trifluoromethyl)benzoic acid

A321785-25G A321785-5G A321785-10G A321785-1G
103825-290EA 9 USD
103825-290 103836-224 76839-136 76839-138 76978-080 77306-252
4-Amino-α,α,α-trifluoro-o-toluic acid 97%
4-Amino-α,α,α-trifluoro-o-toluic acid

Caution: For research use only

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD03407959
CAS Number: 393-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR