(4-Acetamidophenyl)boronic acid 96%
Supplier: Ambeed
|
Warning
|
A195524-1G
A195524-25G
A195524-5G
A195524-10G
A195524-250MG
103816-762EA
8
USD
103816-762
103847-374
77085-326
77085-328
77085-330
77192-138
77192-140
(4-Acetamidophenyl)boronic acid 96%
(4-Acetamidophenyl)boronic acid
Caution: for Research Use Only
Formula:
C₈H₁₀BNO₃ Melting Pt: 215…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179451 CAS Number: 101251-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...