Bathophenanthroline 98%

Supplier: Ambeed
Warning

Synonyms: 4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)

A246584-100G A246584-10G A246584-1MG A246584-250MG A246584-1G A246584-25G A246584-5G A246584-500G
103811-718EA 8 USD
103811-718 103851-552 76786-690 76786-692 76786-694 76786-696 77090-842 77090-844 77090-846 77872-233
Bathophenanthroline 98%
Bathophenanthroline

Caution: for Research Use Only

Formula: C₂₄H₁₆N₂
Boiling Pt: 543.1 °C (760 mm Hg)
Melting Pt: 218…220 °C
Flash Pt: 242.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00004976
CAS Number: 1662-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR