4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one 98%

Supplier: Ambeed
Warning

A111309-100G A111309-10G A111309-25G A111309-500G A111309-1G A111309-5G
103797-814EA 5 USD
103797-814 103823-630 76796-876 76796-878 76796-880 76796-882 77310-432 77310-434
4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one 98%
4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one

Caution: for Research Use Only

Formula: C₁₁H₁₇Cl₂NOS
Boiling Pt: 322.6 °C (760 mm Hg)
Flash Pt: 148.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04034673
CAS Number: 64359-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR