5-Methylfuran-2-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

A402471-1G A402471-25G A402471-5G A402471-100G A402471-10G
103827-076EA 5 USD
103827-076 103838-114 76841-572 76841-574 76841-576 77278-758 77278-760
5-Methylfuran-2-boronic acid pinacol ester 97%
5-Methylfuran-2-boronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₁H₁₇BO₃
Boiling Pt: 277.8 °C (760 mm Hg)
Melting Pt: 217 °C
Storage Temperature: Freezer
MDL Number: MFCD03094689
CAS Number: 338998-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR