(±)-3-Hydroxytetrahydrofuran 98%
Supplier: Ambeed
|
Warning
|
A225792-25G
A225792-5G
A225792-100G
A225792-10G
A225792-500G
103850-100EA
9
USD
103850-100
77088-810
77088-812
77275-622
77275-624
77275-626
(±)-3-Hydroxytetrahydrofuran 98%
(±)-3-Hydroxytetrahydrofuran
Caution: for Research Use Only
Formula:
C₄H₈O₂ Boiling Pt: 181 °C (760 mm Hg) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005374 CAS Number: 453-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...