3-Chlorobenzoic acid 97%

Supplier: Ambeed
Warning

A132909-100G A132909-5G A132909-25G
103808-454EA 65 USD
103808-454 103832-724 76972-188 77298-594 77314-388
3-Chlorobenzoic acid 97%
3-Chlorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 281
Melting Pt: 122
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002491
CAS Number: 535-80-8
Merck Index: 13,02142

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR