3-Chloro-4-((3-fluorobenzyl)oxy)aniline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-4-(3-fluorobenzyloxy)aniline
A266141-25G
A266141-5G
A266141-100G
A266141-10G
A266141-1G
103812-786EA
5
USD
103812-786
103852-686
77092-386
77092-388
77276-690
77276-692
77304-948
3-Chloro-4-((3-fluorobenzyl)oxy)aniline 98%
3-Chloro-4-((3-fluorobenzyl)oxy)aniline
Caution: for Research Use Only
Formula:
C₁₃H₁₁ClFNO Boiling Pt: 387.9 °C (760 mm Hg) Flash Pt: 188.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD06809822 CAS Number: 202197-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...