3-Bromo-2-hydroxypyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromopyridin-2(1H)-one, 3-Bromopyridin-2-ol, 3-Bromo-2-pyridinol
A604744-1G
A604744-25G
A604744-5G
A604744-100G
A604744-10G
A604744-500G
103828-994EA
5
USD
103828-994
103840-136
76844-294
76844-296
76844-298
77157-634
77157-636
77686-530
3-Bromo-2-hydroxypyridine 98%
3-Bromo-2-hydroxypyridine
Caution: for Research Use Only
Formula:
C₅H₄BrNO MW: 174 g/mol Melting Pt: 179…183 °C Flash Pt: 157.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411569 CAS Number: 13466-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...