3-Amino-3-phenyl-1-propanol 97%
Supplier: Ambeed
|
Danger
|
A105017-100G
A105017-5G
103796-814EA
6
USD
103796-814
103822-542
77148-400
77283-332
3-Amino-3-phenyl-1-propanol 97%
3-Amino-3-phenyl-1-propanol
Caution: for Research Use Only
Formula:
C₉H₁₃NO Boiling Pt: 293 °C (760 mm Hg) Melting Pt: 70…77 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00801124 CAS Number: 14593-04-5 UN: 3259 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...