Myricetin 98%

Supplier: Ambeed
Warning

Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

A268149-10G A268149-1G A268149-5G A268149-50MG A268149-100G A268149-1MG
103812-898EA 11 USD
103812-898 103852-800 76722-982 77305-000 77305-002 77593-656 77668-162 77668-164
Myricetin 98%
Myricetin

Caution: for Research Use Only

Formula: C₁₅H₁₀O₈
MW: 318.24 g/mol
Melting Pt: >300 °C
Flash Pt: 285.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00006827
CAS Number: 529-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR