3,4-Dimethylbenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A238431-25G
A238431-500G
A238431-5G
A238431-100G
103809-680EA
65
USD
103809-680
77090-130
77275-956
77275-958
77568-304
3,4-Dimethylbenzaldehyde 98%
3,4-Dimethylbenzaldehyde
Caution: For research use only.
Formula:
C₉H₁₀O Boiling Pt: 226 °C (760 mm Hg) Melting Pt: 225…226 °C Flash Pt: 97.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016612 CAS Number: 5973-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...