3,4-Dimethoxyaniline 98%

Supplier: Ambeed
Warning

A173353-100G A173353-10G A173353-500G A173353-25G A173353-1000G
103845-106EA 6 USD
103845-106 76813-284 76813-286 76813-288 77301-260 77611-690
3,4-Dimethoxyaniline 98%
3,4-Dimethoxyaniline

Caution: for Research Use Only

Formula: C₈H₁₁NO₂
Boiling Pt: 296.3 °C (760 mm Hg)
Melting Pt: 85…89 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008394
CAS Number: 6315-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR