Tetrafluorophthalonitrile 97%

Supplier: Ambeed
Warning

A134254-10G A134254-500G A134254-5G A134254-1G A134254-25G
103801-990EA 7 USD
103801-990 103832-934 76782-548 76782-550 77298-702 77314-724 77314-726
Tetrafluorophthalonitrile 97%
Tetrafluorophthalonitrile

Caution: for Research Use Only

Formula: C₈F₄N₂
Boiling Pt: 274.2 °C (760 mm Hg)
Melting Pt: 81…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00001774
CAS Number: 1835-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR