2-Hydroxynicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxypyridine-3-carboxylic acid, 1,2-Dihydro-2-oxonicotinic acid, 2-Hydroxy-3-pyridinecarboxylic acid, 2-Oxo-1,2-dihydropyridine-3-carboxylic acid

A120332-100G A120332-10G A120332-500G A120332-1000G A120332-25G
103808-218EA 147 USD
103808-218 77108-530 77108-532 77108-534 77108-536 77312-024 77685-057
2-Hydroxynicotinic acid 98%
2-Hydroxynicotinic acid

Caution: for Research Use Only

Formula: C₆H₅NO₃
Boiling Pt: 436 °C (760 mm Hg)
Melting Pt: 258…261 °C
Flash Pt: 217.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00010100
CAS Number: 609-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR