2-Methoxy-5-methylphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methoxy-5-methylbenzeneboronic acid
A131966-10G
A131966-5G
103801-668EA
5
USD
103801-668
103832-574
77190-090
77298-528
77314-204
2-Methoxy-5-methylphenylboronic acid 97%
2-Methoxy-5-methylphenylboronic acid
Caution: for Research Use Only
Formula:
C₈H₁₁BO₃ MW: 165.99 g/mol Boiling Pt: 342 °C (760 mm Hg) Melting Pt: 92…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179465 CAS Number: 127972-00-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...