2-Fluoro-5-nitrophenol 98%

Supplier: Ambeed
Danger

A348054-1G A348054-100G A348054-10G A348054-250MG
103825-986EA 9 USD
103825-986 103836-972 76840-048 77263-822 77263-824 77263-826
2-Fluoro-5-nitrophenol 98%
2-Fluoro-5-nitrophenol

Caution: for Research Use Only

Formula: C₆H₄FNO₃
MW: 157.1000 g/mol
Boiling Pt: 256.7 °C (760 mm Hg)
Flash Pt: 109.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09054718
CAS Number: 22510-08-3
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR