2-Fluoro-5-(trifluoromethyl)pyridine 96%
Supplier: Ambeed
|
Danger
|
A631690-5G
A631690-10G
A631690-25G
103840-274EA
6
USD
103840-274
76844-442
77266-974
77308-012
77308-014
2-Fluoro-5-(trifluoromethyl)pyridine 96%
2-Fluoro-5-(trifluoromethyl)pyridine
Caution: for Research Use Only
Formula:
C₆H₃F₄N Boiling Pt: 135.3 °C (760 mm Hg) Flash Pt: 32.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07437945 CAS Number: 69045-82-5 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...