2-Fluoro-3-nitropyridine 98%

Supplier: Ambeed
Warning

A128491-100G A128491-10G A128491-1G A128491-500G A128491-25G A128491-5G
103832-020EA 18 USD
103832-020 76996-216 76996-218 76996-220 76996-222 77313-464 77313-466
2-Fluoro-3-nitropyridine 98%
2-Fluoro-3-nitropyridine

Caution: for Research Use Only

Formula: C₅H₃FN₂O₂
MW: 142.09
Boiling Pt: 248 °C (760 mm Hg)
Flash Pt: 103.842 °C
Storage Temperature: Ambient
MDL Number: MFCD03095068
CAS Number: 1480-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR