2-Chlorothiazole 98%

Supplier: Ambeed
Danger

A169092-25G A169092-10G A169092-5G
103844-608EA 5 USD
103844-608 77081-680 77163-464 77300-960
2-Chlorothiazole 98%
2-Chlorothiazole

Caution: for Research Use Only

Formula: C₃H₂ClNS
Boiling Pt: 180.8 °C (760 mm Hg)
Flash Pt: 54.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00210701
CAS Number: 3034-52-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR