(2-Chlorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chlorophenylacetic acid

A326510-100G A326510-500G A326510-25G
103810-134EA 209 USD
103810-134 77239-678 77239-680 77306-348
(2-Chlorophenyl)acetic acid 98%
(2-Chlorophenyl)acetic acid

Caution: for Research Use Only

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 294.1 °C (760 mm Hg)
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00004317
CAS Number: 2444-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR