2-chloro-5-iodopyrimidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-5-iodopyrimidine
A128084-100G
A128084-10G
A128084-250MG
A128084-1G
103801-064EA
7
USD
103801-064
103831-940
76971-946
76971-948
76971-950
77313-350
2-chloro-5-iodopyrimidine 98%
2-chloro-5-iodopyrimidine
Caution: for Research Use Only
Formula:
C₄H₂ClIN₂ Boiling Pt: 317.8 °C (760 mm Hg) Melting Pt: 128…131 °C Flash Pt: 146 °C Storage Temperature: Freezer |
MDL Number:
MFCD06200210 CAS Number: 32779-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...