2-Chloro-5-(trifluoromethyl)benzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A270972-5G
A270972-100G
103852-928EA
14
USD
103852-928
77092-700
77194-010
77194-012
2-Chloro-5-(trifluoromethyl)benzaldehyde 98%
2-Chloro-5-(trifluoromethyl)benzaldehyde
Caution: for Research Use Only
Formula:
C₈H₄ClF₃O Boiling Pt: 42…44 °C (1.5 mm Hg) Flash Pt: 89.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00134430 CAS Number: 82386-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...