2-Bromethylamine hydrobromide 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

A156289-100G A156289-500G A156289-1000G A156289-25G
103808-830EA 108 USD
103808-830 76997-538 76997-540 77318-816 77318-818
2-Bromethylamine hydrobromide 97%
2-Bromethylamine hydrobromide

Caution: for Research Use Only

Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012886
CAS Number: 2576-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR