2-Bromo-5-chloropyridine 98%
Supplier: Ambeed
|
Warning
|
A114272-100G
A114272-10G
A114272-1G
A114272-25G
103808-120EA
297
USD
103808-120
103824-222
76982-882
76982-884
76982-886
76982-888
2-Bromo-5-chloropyridine 98%
2-Bromo-5-chloropyridine
Caution: for Research Use Only
Formula:
C₅H₃BrClN Boiling Pt: 212 °C (760 mm Hg) Melting Pt: 65…69 °C Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234006 CAS Number: 40473-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...