2-Amino-6-chloropyridine 98%

Supplier: Ambeed
Warning

A175380-100G A175380-10G A175380-500G A175380-25G A175380-5G
103845-342EA 5 USD
103845-342 77011-732 77011-734 77011-736 77082-686 77301-370
2-Amino-6-chloropyridine 98%
2-Amino-6-chloropyridine

Caution: for Research Use Only

Formula: C₅H₅ClN₂
Boiling Pt: 255.7 °C (760 mm Hg)
Melting Pt: 69…73 °C
Flash Pt: 108.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00234068
CAS Number: 45644-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR