2-Aminonicotinonitrile 96%

Supplier: Ambeed
Warning

Synonyms: 2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine

A247400-25G A247400-5G A247400-1G
103811-766EA 10 USD
103811-766 103851-600 77090-936 77090-938 77304-404
2-Aminonicotinonitrile 96%
2-Aminonicotinonitrile

Caution: for Research Use Only

Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 297.6 °C (760 mm Hg)
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00128867
CAS Number: 24517-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR