2,6-Dichloropurine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloro-9H-purine

A101242-100G A101242-10G A101242-25G A101242-500G A101242-5G
103795-868EA 5 USD
103795-868 103821-552 77270-212 77270-214 77270-216 77270-218 77282-686
2,6-Dichloropurine 98%
2,6-Dichloropurine

Caution: for Research Use Only

Formula: C₅H₂Cl₂N₄
MW: 189 g/mol
Boiling Pt: 405.4 °C (760 mm Hg)
Melting Pt: 181
Flash Pt: 231.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00077725
CAS Number: 5451-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR