2,4-Dichloro-5-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-5-picoline

A183924-100MG A183924-1G A183924-25G A183924-5G A183924-10G
103815-686EA 8 USD
103815-686 103846-248 76974-540 77083-858 77083-860 77083-862 77611-788
2,4-Dichloro-5-methylpyridine 98%
2,4-Dichloro-5-methylpyridine

Caution: for Research Use Only

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 221.2 °C (760 mm Hg)
Flash Pt: 108.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08704813
CAS Number: 56961-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR