2,4-Dichloro-5-nitropyrimidine 97% (contains ≤10%DCM)

Supplier: Ambeed
Warning

A190186-10G A190186-25G A190186-100G
103846-872EA 5 USD
103846-872 76825-244 77084-660 77550-212
2,4-Dichloro-5-nitropyrimidine 97% (contains ≤10%DCM)
2,4-Dichloro-5-nitropyrimidine

Caution: for Research Use Only

Formula: C₄HCl₂N₃O₂
Boiling Pt: 325.8 °C (760 mm Hg)
Melting Pt: 28…32 °C
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00127867
CAS Number: 49845-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR