2,3,5-Trifluoropyridine 98%
Supplier: Ambeed
|
Danger
|
A667946-1G
A667946-5G
A667946-25G
103840-484EA
27
USD
103840-484
76844-688
76844-690
77573-154
2,3,5-Trifluoropyridine 98%
2,3,5-Trifluoropyridine
Caution: for Research Use Only
Formula:
C₅H₂F₃N Boiling Pt: 102 °C (760 mm Hg) Flash Pt: 30 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03001162 CAS Number: 76469-41-5 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...