2,3,4,5-Tetrahydro-1H-benzo[b]azepine 96%
Supplier: Ambeed
|
Warning
|
A165445-100MG
A165445-250MG
A165445-1G
A165445-5G
103805-970EA
14
USD
103805-970
103844-224
77011-374
77011-376
77300-788
77320-434
2,3,4,5-Tetrahydro-1H-benzo[b]azepine 96%
2,3,4,5-Tetrahydro-1H-benzo[b]azepine
Caution: for Research Use Only
Formula:
C₁₀H₁₃N Boiling Pt: 268.9 °C (760 mm Hg) Melting Pt: 30…34 °C Flash Pt: 122.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272363 CAS Number: 1701-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...