PIPES 98%

Supplier: Ambeed
Warning

Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), Piperazine-1,4-bis(2-ethanesulfonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)diethanesulfonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid, Piperazine-N,N'-bis-2-ethanesulfonic acid

A145800-100G A145800-10G A145800-500G A145800-25G
103808-668EA 129 USD
103808-668 77027-884 77027-886 77027-888 77299-476
PIPES 98%
PIPES

Caution: for Research Use Only

Formula: C₈H₁₈N₂O₆S₂
MW: 302.37 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006159
CAS Number: 5625-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR