1-Nitro-4-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethoxy)nitrobenzene

A217495-10G A217495-5G
103849-352EA 6 USD
103849-352 76814-804 77087-976
1-Nitro-4-(trifluoromethoxy)benzene 98%
1-Nitro-4-(trifluoromethoxy)benzene

Caution: for Research Use Only

Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Boiling Pt: 217.4 °C (760 mm Hg)
Flash Pt: 85.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00040302
CAS Number: 713-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR