1H-Pyrazole-1-carboximidamide hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine
A107913-100G
A107913-10G
A107913-25G
A107913-500G
A107913-5G
103807-980EA
337
USD
103807-980
103823-036
77233-290
77233-292
77233-294
77233-296
77309-882
1H-Pyrazole-1-carboximidamide hydrochloride 97%
1H-Pyrazole-1-carboximidamide hydrochloride
Caution: for Research Use Only
Formula:
C₄H₇ClN₄ MW: 146.58 g/mol Melting Pt: 7…170 °C Flash Pt: 493 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210087 CAS Number: 4023-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...