1,2-Difluoro-4-iodobenzene 95% stabilised

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluoroiodobenzene

Stabilization: Stabilised with copper chip

A144071-100G A144071-10G A144071-1G A144071-5G A144071-25G
103841-436EA 9 USD
103841-436 77285-858 77285-860 77285-862 77299-364 77316-676
1,2-Difluoro-4-iodobenzene 95% stabilised
1,2-Difluoro-4-iodobenzene

Caution: for Research Use Only

Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 177
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044630
CAS Number: 64248-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR