1,2-Bis(diphenylphosphino)ethane nickel(II) chloride 98%

Supplier: Ambeed
Warning

Synonyms: Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)

A100654-100G A100654-25G A100654-500G A100654-1G A100654-5G
103795-552EA 5 USD
103795-552 103821-248 76994-456 76994-458 76994-460 77282-532 77296-784
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride 98%
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride

Caution: for Research Use Only

Formula: C₂₆H₂₄Cl₂NiP₂
Melting Pt: 263…265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013313
CAS Number: 14647-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR