1,10-Phenanthroline monohydrate 98%

Supplier: Ambeed
Danger

Synonyms: 1,10-Phenanthroline hydrate

A255329-100G A255329-10G A255329-25MG A255329-500G A255329-5G A255329-25G
103809-814EA 330 USD
103809-814 103852-044 76976-786 76976-788 76976-790 76976-792 76976-794 77091-506
1,10-Phenanthroline monohydrate 98%
1,10-Phenanthroline monohydrate

Caution: for Research Use Only

Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Boiling Pt: 365.1 °C (760 mm Hg)
Melting Pt: 100…104 °C
Flash Pt: 164.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR