1-(p-Toluenesulfonyl)imidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(P-Toluenesulfonyl)imidazole, 1-(P-Toluenesulphonyl)imidazole, 1-Tosylimidazole

A116513-100G A116513-10G A116513-25G A116513-5G A116513-500G A116513-1000G
103824-610EA 5 USD
103824-610 77189-408 77189-410 77189-412 77311-418 77614-916 77745-859
1-(p-Toluenesulfonyl)imidazole 98%
1-(p-Toluenesulphonyl)imidazole

Caution: for Research Use Only

Formula: C₁₀H₁₀N₂O₂S
MW: 222.26 g/mol
Boiling Pt: 409.1 °C (760 mm Hg)
Melting Pt: 76…78 °C
Flash Pt: 201.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00005285
CAS Number: 2232-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR