4'-Hydroxychalcone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Hydroxyphenyl)-3-phenylprop-2-en-1-one, 4'-Hydroxybenzylidenacetophenone

A131746-100G A131746-100MG A131746-10G A131746-1G A131746-25G A131746-5G
103801-622EA 9 USD
103801-622 103832-530 76870-774 76870-776 76870-778 77298-506 77314-132 77314-134
4'-Hydroxychalcone 97%
4'-Hydroxychalcone

Caution: for Research Use Only

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Boiling Pt: 419.6 °C (760 mm Hg)
Melting Pt: 174…178 °C
Flash Pt: 179.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00016484
CAS Number: 2657-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR