(S)-(+)-α-Methoxyphenylacetic acid 98%
Supplier: Ambeed
|
Warning
|
A791171-1G
A791171-5G
A791171-100G
A791171-10G
A791171-250MG
A791171-25G
103841-020EA
33
USD
103841-020
76845-268
76845-270
77255-878
77255-880
77255-882
77255-884
(S)-(+)-α-Methoxyphenylacetic acid 98%
(S)-(+)-α-Methoxyphenylacetic acid
Caution: for Research Use Only
Formula:
C₉H₁₀O₃ Boiling Pt: 283.8 °C (760 mm Hg) Melting Pt: 64…66 °C Flash Pt: 116.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064216 CAS Number: 26164-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...