(S)-(-)-4-Fluoro-α-methylbenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (S)-1-(4-Fluorophenyl)ethanamine, (S)-(-)-1-(4-Fluorophenyl)ethylamine

A185149-10G A185149-1G A185149-25G A185149-5G
103815-824EA 8 USD
103815-824 103846-392 77111-558 77301-950 77301-952 77301-954
(S)-(-)-4-Fluoro-α-methylbenzylamine 98%
(S)-(-)-4-Fluoro-α-methylbenzylamine

Caution: for Research Use Only

Formula: C₈H₁₀FN
Boiling Pt: 185.4 °C (760 mm Hg)
Melting Pt: …30 °C
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092999
CAS Number: 66399-30-2
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR