(R)-(-)-1,2-Propanediol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-1,2-Dihydroxy propane, (R)-(-)-1,2-Propylene glykol, (R)-Propane-1,2-diol

A289494-25G A289494-5G A289494-10G A289494-100G A289494-500G A289494-1000G
103813-834EA 5 USD
103813-834 103834-974 76837-552 76837-554 77141-312 77616-250 77616-252 77746-535
(R)-(-)-1,2-Propanediol 98%
(R)-(-)-1,2-Propanediol

Caution: for Research Use Only

Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 184.8 °C (760 mm Hg)
Melting Pt: 57 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00066248
CAS Number: 4254-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR