(R)-3-Hydroxybutyric acid 95%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Hydroxybutanoic acid, (R)-beta-Hydroxybutyric acid

A257416-1G A257416-5G A257416-10G A257416-250MG A257416-100G A257416-25G
103799-532EA 37 USD
103799-532 103812-304 103852-172 77091-678 77091-680 77140-660 77140-662 77568-488 77746-417
(R)-3-Hydroxybutyric acid 95%
(R)-3-Hydroxybutyric acid

Caution: For research use only.

Formula: C₄H₈O₃
Melting Pt: 49…50 °C
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066257
CAS Number: 625-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR