(R)-(-)-2-Oxothiazolidine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A144428-100G A144428-10G A144428-25G A144428-1G A144428-5G A144428-500G
103803-402EA 6 USD
103803-402 103841-478 76997-000 76997-002 77299-380 77316-716 77316-718 77671-048
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid 98%
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid

Caution: for Research Use Only

Formula: C₄H₅NO₃S
Melting Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00066092
CAS Number: 19771-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR