Resveratrol 98%

Supplier: Ambeed
Warning

Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol

A419526-5G A419526-10G A419526-25G A419526-100G A419526-500G
103810-314EA 477 USD
103810-314 103838-416 76842-032 77143-136 77143-138 77307-202 77568-846 77746-841
Resveratrol 98%
Resveratrol

Caution: For research use only.

Formula: C₁₄H₁₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00133799
CAS Number: 501-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR