3-Chloro-2-fluorophenylhydrazine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-2-fluorophenyl)hydrazine hydrochloride

A159046-250MG A159046-1G A159046-5G
103805-218EA 10 USD
103805-218 103843-430 77286-390 77319-298 77319-300
3-Chloro-2-fluorophenylhydrazine hydrochloride 97%
3-Chloro-2-fluorophenylhydrazine hydrochloride

Caution: for Research Use Only

Formula: C₆H₇Cl₂FN₂
MW: 197.0400 g/mol
Boiling Pt: 235.1 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094280
CAS Number: 517920-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR